Detergent, Contec® Sterile NeutraKlean
供应商: CONTEC
SBC503NK SBT103NK
CONTSBC503NKEA 0 CNY
CONTSBC503NK CONTSBT103NK
Detergent, Contec® Sterile NeutraKlean
清洗剂 洗涤剂和表面清洁剂
Contec® Sterile NeutraKlean is a gentle and low-foaming detergent designed for the cleaning of life science cleanrooms. Ready to use, irradiated neutral detergent with purified water.
  • Neutral pH makes it suitable for use in all areas of cleanroom as gentle on surfaces
  • Filtered to 0.2 microns under Grade A airflow in an Grade C cleanroom ensures the product is free from contamination and particulates
  • Low-foaming detergent action is ideal for use on floors, walls, and benches
  • Low level of surfactant provides low residue which can be easily removed
  • Trigger
More Product Information