Cleanroom wipes, pre-saturated, sterile, PROSAT® Sterile Sigma
供应商: CONTEC
PSC20003
CONTPSC20003EA 0 CNY
CONTPSC20003
Cleanroom wipes, pre-saturated, sterile, PROSAT® Sterile Sigma
擦拭巾 洁净室擦拭巾 Presaturated Cleanroom Wipes
These PROSAT® Sterile wipes are manufactured from hydroentangled cellulose/polyester and saturated with a solution of 70% denatured ethanol and 30% purified water.
  • Non woven material is low in particles and fibres making an excellent general purpose wipes
  • Sterile per gamma irradiation so suitable for Grade A/B environments
  • Highly sorbent with good wet strength
  • Pre-saturated wipe reduces VOC emissions and solvent use
  • Resealable pouch preserves cleanliness and solvent saturation levels
  • Increases process control and repeatability
A cost effective wipeMore Product Information