Cleanroom wipes, Polynit
供应商: CONTEC
PN-99 LWPN0003
CONTPN-99EA 0 CNY
CONTPN-99 CONTLWPN0003
Cleanroom wipes, Polynit
擦拭巾 洁净室擦拭巾 Dry Cleanroom Wipes
Polynit wipes are made of 100% knitted textured polyester with knife cut edges for a very soft wipe.
  • Laundered knitted 100% polyester fabric for extremely low levels of particles and fibres
  • Soft edge is ideal for use on surfaces which are vulnerable to scratching
  • 100% knitted polyester is compatible with many solvents
  • Durable fabric is abrasion and chemical resistant
  • Laundered and packaged in an ISO Class 4 cleanroom
An interlock knit creates a durable fabric. The addition of a periodic additional stitch of a no-run interlock knit prevents the fabricMore Product Information