Mopping systems, EasyCurve™
供应商: CONTEC
EZSE0100
CONTEZSE0100EA 0 CNY
CONTEZSE0100
Mopping systems, EasyCurve™
拖把
Contec’s EasyCurve™ Mop was designed specifically for critical and controlled environments, now available with a ribbed microfiber face giving users smooth and easy mopping for better particle pick up and contamination removal. The combination of the curved mop face and the pivoting connector results in effortless surface contact and outstanding maneuverability, making the EasyCurve™ an ideal cleaning tool for ceilings, walls and floors. Available in nonsterile and validated sterile versions.
Mop heads:
Very absorbent and drip-free
Compatible with most common solutions, solvents andMore Product Information