Tin(II) chloride

供应商: LOCAL DEALS
Danger

Synonyms: Stannous chloride, Tin dichloride

R02061710124
CHLCR02061710124EA 0 CNY
CHLCR02061710124
Tin(II) chloride
Tin(II) chloride
分子式: SnCl₂
分子量: 189.62 g/mol
沸点: 623
溶点: 246
密度: 3.95 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00011241
CAS号: 7772-99-8
EINECS: 231-868-0
UN号: 3260
ADR: 8 III
Merck Index: 13,08861

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR