2-氨基-6-氯吡啶

供应商: LOCAL DEALS
Warning

Synonyms: 6-氯-2-吡啶胺

R01825130131
CHLCR01825130131EA 0 CNY
CHLCR01825130131
2-氨基-6-氯吡啶
2-氨基-6-氯吡啶
分子式: C₅H₅ClN₂
分子量: 128.56 g/mol
MDL编号: MFCD00234068
CAS号: 45644-21-1

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR