1,4-二氧六环

供应商: LOCAL DEALS
Danger

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

R00273320024 R00273320024CD
CHLCR00273320024EA 0 CNY
CHLCR00273320024 CHLCR00273320024CD
1,4-二氧六环
1,4-二氧六环
分子式: C₄H₈O₂
分子量: 88.11 g/mol
沸点: 101.1
溶点: 11.8
密度: 1.03 g/cm³ (20 °C)
闪点: 12
存储条件: Ambient
MDL编号: MFCD00006571
CAS号: 123-91-1
EINECS: 204-661-8
UN号: 1165
ADR: 3 II
Merck Index: 13,03330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR