Stirrer bearings, PTFE
供应商: CHEMGLASS
CG-2077-20 CG-2077-21 CG-2077-01 CG-2077-10
CGLACG-2077-20EA 0 CNY
CGLACG-2077-20 CGLACG-2077-21 CGLACG-2077-01 CGLACG-2077-10
Stirrer bearings, PTFE
搅拌器 Stirring Tools
These grease-free PTFE stirrer bearings consist of a threaded glass adapter with the listed inner joint, a saddle o-ring, a body bushing with a compression and locking nut, and a precision PTFE inner bearing.
  • Ideal for use in moderate vacuum applications
  • Saddle o-ring allows only glass and PTFE contact with the reaction, while ensuring a vacuum tight seal