Connecting adapters, for rotary evaporators to vials
供应商: CHEMGLASS
CG-1318-30 CG-1318-12 CG-1318-25 CG-1318-10 CG-1318-24 CG-1318-40 CG-1318-22
CGLACG-1318-30EA 0 CNY
CGLACG-1318-30 CGLACG-1318-12 CGLACG-1318-25 CGLACG-1318-10 CGLACG-1318-24 CGLACG-1318-40 CGLACG-1318-22
Connecting adapters, for rotary evaporators to vials
适配器 带连接管接头的适配器
These adapters permit the connection of standard laboratory dram sample vials directly to the vapor tube or a bump trap of a rotary evaporator.
  • Very good chemical resistance
  • Very good temperature resistance
With a top outer joint of the listed size and a 20 to 400 G.P.I. thread at the bottom. A PTFE lined, polypropylene double-threaded connector attaches the vial to the adapter.
订购须知 The glass adapter and double-threaded connections can be ordered separately.
Delivery information: Complete kits includes glass adapter and the 6 sizes of theMore Product Information