Dipping bottle, 'Ex'
供应商: BURKLE
5365-3000
BURK5365-3000EA 0 CNY
BURK5365-3000
Dipping bottle, 'Ex'
采样器 样条
Dipping bottle for testing mineral oil products (Category A, Explosion Categories IIA, IIB and IIC), crude oil, liquid mineral oil products, fuels, special and test fuels, fuel oils, petroleum and liquid lubricants. Completely made from non spark-producing materials, nickel plated.
  • Screw-off bottle head
  • Easy to clean robust handle
  • High weight (3,2 kg) for fast lowering
Certifications: Tested for use in the Ex protection zone 0.
订购须知 Please order the appropriate lowering cable at the same time.