Hand drill, QualiRod, cylindrical
供应商: BURKLE
5311-0101
BURK5311-0101EA 0 CNY
BURK5311-0101
Hand drill, QualiRod, cylindrical
采样器 固体取样器
Cylindrical hollow drill with a sharp cutting edge, can easily be driven into paste-like and semi-solid substances. The samples cut out are subsequently removed with the appropriate push-rod ejector.
  • Stainless steel V2A (1,4301)
  • Sterilisable
  • Inner Ø 8 mm, outer Ø 10 mm