Control seals, close-it food & pharma
供应商: BURKLE
5303-2017 5303-2018 5303-3018 5303-3017 5303-2020 5303-2023 5303-2021 5303-2022 5303-3020 5303-3022 5303-3021 5303-3023
BURK5303-2017EA 0 CNY
BURK5303-2017 BURK5303-2018 BURK5303-3018 BURK5303-3017 BURK5303-2020 BURK5303-2023 BURK5303-2021 BURK5303-2022 BURK5303-3020 BURK5303-3022 BURK5303-3021 BURK5303-3023
Control seals, close-it food & pharma
标签
A control seal with an adhesive approved for use with foodstuffs. The strength of the adhesive is almost as powerful as that of the original. Just like the original, close-it food and pharma has a barrier layer of aluminium foil.
  • Adheres very firmly
  • Can be written on
  • Adhesive complies with the FDA and European guidelines for foodstuffs
Suitable for sampling foodstuffs, pharmaceuticals, cosmetics, food additives, even frozen goods where cleanliness is particularly important.