Volumetric flask, class A, glass stopper with individual certificate, BLAUBRAND®
供应商: BRAND
937266
BRND937266EA 0 CNY
BRND937266
Volumetric flask, class A, glass stopper with individual certificate, BLAUBRAND®
烧瓶 容量瓶
Borosilicate glass 3.3, with glass stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • DE-M marking
  • Individual certificate included