384-well microplates for immunology, BRANDplates®
供应商: BRAND
781740 781741 781742
BRND781740EA 0 CNY
BRND781740 BRND781741 BRND781742
384-well microplates for immunology, BRANDplates®
微孔板 免疫微孔板和条带
PS, solid or with transparent base. These flat bottom plates are available in different colours, and with standard or transparent base formats.
immunoGrade™: The surface of these plates is optimised for the immobilisation of IgG, showing a high binding capacity for molecules with hydrophilic and hydrophobic regions. Plates with an immunoGrade™ surface are the best choice for standard ELISA applications.
The surface of these plates is optimised for the immobilisation of IgG, showing a high binding capacity for molecules with hydrophilic and hydrophobic regions. Plates with anMore Product Information