Funnel stands
供应商: BRAND
74511 74506
BRND74511EA 0 CNY
BRND74511 BRND74506
Funnel stands
支撑架
PP plates for holding two or four funnels with top outer-Ø of 50 to 120 mm.
  • With aluminum support rod (Ø×L: 12,7×595 mm)
  • Adjustable funnel height