Density bottles, uncalibrated
供应商: BRAND
43205 43208 43220 43228 43238
BRND43205EA 0 CNY
BRND43205 BRND43208 BRND43220 BRND43228 BRND43238
Density bottles, uncalibrated
比重瓶
Borosilicate 3.3 glass, Gay-Lussac type
  • Stopper NS 10/19 with capillary
  • Top of stopper ground and polished
  • Nominal capacity printed on the bottom
Certifications: DIN ISO 3507