Volumetric flasks, class A, white markings, BLAUBRAND®
供应商: BRAND
37438 37443 37445 37446 37447 37448 37449 37450 37451 37452 37453
BRND37438EA 0 CNY
BRND37438 BRND37443 BRND37445 BRND37446 BRND37447 BRND37448 BRND37449 BRND37450 BRND37451 BRND37452 BRND37453
Volumetric flasks, class A, white markings, BLAUBRAND®
烧瓶 容量瓶
Borosilicate glass 3.3, amber, with glass stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast white enamel
  • DE-M marking
*Wide neck.
Certifications: DIN EN ISO 1042.
订购须知 Supplied with a batch certificate. Volumetric flasks also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.