b.safe Funnels 180
供应商: BOHLENDER
R800-02 R800-05 R800-10 R801-30 R810-05 R811-30 R820-05 R821-30 R830-05 R830-10 R831-30 R840-05 R840-10 R841-30 R850-05 R851-30 R860-05 R861-30 R870-05 R871-30 R880-05 R880-10 R890-05 R890-10
BOHLR800-02EA 0 CNY
BOHLR800-02 BOHLR800-05 BOHLR800-10 BOHLR801-30 BOHLR810-05 BOHLR811-30 BOHLR820-05 BOHLR821-30 BOHLR830-05 BOHLR830-10 BOHLR831-30 BOHLR840-05 BOHLR840-10 BOHLR841-30 BOHLR850-05 BOHLR851-30 BOHLR860-05 BOHLR861-30 BOHLR870-05 BOHLR871-30 BOHLR880-05 BOHLR880-10 BOHLR890-05 BOHLR890-10
b.safe Funnels 180
漏斗 安全漏斗
Funnel made of PE / static dissipative PE-EX for liquid waste disposal. The funnel can be safely mounted on the waste container with its freely turnable cap that is safely fixed with the filling neck.
  • Easy and safe handling: you do not need to open a lid before pouring
  • Removable sieve restrains, e.g. magnetic stirring bars or other solids
  • Splash guard for a smooth flow without splashing
  • Ball valve protects from harmful vapours; it floats up during filling and opens the drain
  • Free turnable cap for easy assembly on collecting container
  • Neck with plastic nibs
More Product Information