Caps for solvent supplies, b.safe
供应商: BOHLENDER
M128-01 M128-02 M138-01 M138-02 M138-03 M139-01 M139-02 M139-03 M140-01 M140-02 M140-03 M141-01 M143-01 M143-02 M143-03 M144-01 M144-02 M144-03 M144-05 M145-01 M145-02 M145-03 M145-04 M145-05 M147-01 M147-02 M180-01 M180-02 M180-03 M183-04 M529-01
BOHLM128-01EA 0 CNY
BOHLM128-01 BOHLM128-02 BOHLM138-01 BOHLM138-02 BOHLM138-03 BOHLM139-01 BOHLM139-02 BOHLM139-03 BOHLM140-01 BOHLM140-02 BOHLM140-03 BOHLM141-01 BOHLM143-01 BOHLM143-02 BOHLM143-03 BOHLM144-01 BOHLM144-02 BOHLM144-03 BOHLM144-05 BOHLM145-01 BOHLM145-02 BOHLM145-03 BOHLM145-04 BOHLM145-05 BOHLM147-01 BOHLM147-02 BOHLM180-01 BOHLM180-02 BOHLM180-03 BOHLM183-04 BOHLM529-01
Caps for solvent supplies, b.safe
外壳 安全塞和废品塞
b.safe caps together with fittings, tubing and an air valve form a perfect unit for solvent extraction from laboratory flasks.
  • Perfect protection from hazardous vapours
  • Closed system: reduction of expensive solvent loss
  • Guarantee of pure eluents
  • Huge choice of cap sizes for compatibility with any eluent flask
  • No twisting of capillaries
  • Screw caps can individually be labelled by hand or with removable, self-adhesive labels
b.safe air valve: The special valve lets flow into the bottle as much ambient air as necessary for pressure compensation duringMore Product Information