Sample tubes for stool, with screw cap
供应商: BOETTGER PAUL
07.033.3107
BOET07.033.3107EA 0 CNY
BOET07.033.3107
Sample tubes for stool, with screw cap
临床诊断系统 Clinical Sample Collection/Preparation Faeces Sample Collection
PP, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
Clair tube with overlapping screw cap for optimal hygiene when opening, reliably sealed with long-lasting spoon for about bean-sized samples.
Certifications: Conforms to DIN EN 829 for the dispatch of stool samples.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904