Tissue float baths, deluxe
供应商: BOEKEL SCIENTIFIC
145951-2
BOEI145951-2EA 0 CNY
BOEI145951-2
Tissue float baths, deluxe
浴锅 组织浴
Precision temperature controlled water bath that eliminates wrinkles and distortion during the preparation of paraffin embedded tissue sections.
  • Digital microprocessor control
  • LED with illuminated black background, transparent PYREX® dish allows specimen to be easily seen
  • Slide dryer block to assist in drying slides
  • Tissue orienter block to smooth wrinkled sections
现在订购
规格
Environmentally Preferable
温度范围 5...65 °C
准确度 ±1 °C @ 55 °C
电气 230 V, 50/60 Hz
外部宽度×深度×高度 362×362×114 mm
Weight 4,7 kg