Stirrers, propeller, with two paddles
供应商: BOCHEM
9605 9604 9603
BOCH9605EA 0 CNY
BOCH9605 BOCH9604 BOCH9603
Stirrers, propeller, with two paddles
搅拌器 Stirring Tools
Propeller stirrer made of stainless steel with two flexible paddles.
  • Mixing of media with low and middle viscosity
  • Radial flow