Overhead stirrer tools, paddle stirrers
供应商: BOCHEM
9503 9507 9504 9505 9506 9508
BOCH9503EA 0 CNY
BOCH9503 BOCH9507 BOCH9504 BOCH9505 BOCH9506 BOCH9508
Overhead stirrer tools, paddle stirrers
搅拌器 Stirring Tools
Paddle stirrers made of stainless steel, with six holes.
  • Ideal for stirring liquids at middle speed range
  • Suitable for stirring low to medium viscous media
  • Creates radial flow