Pharma Scoops
供应商: BOCHEM
8860 8861 8862 8863
BOCH8860EA 0 CNY
BOCH8860 BOCH8861 BOCH8862 BOCH8863
Pharma Scoops
样品铲
These scoops are made from V4A steel.
  • Very good chemical resistance
  • Excellent thermal resistance, autoclavable
  • Highly polished
  • With seamless fixed handle
Ideal for sample taking in the pharmaceutical industry.