Burette and thermometer clamps with bosshead, AUTOMATIC
供应商: BOCHEM
5625
BOCH5625EA 0 CNY
BOCH5625
Burette and thermometer clamps with bosshead, AUTOMATIC
夹具 通用夹具
These chromium plated clamps are designed for holding burettes or thermometers (Ø 0 - 25 mm). The bosshead (Ø 13,5 mm) can be rotated and locked at an angle of 90°.