Laboratory jacks
供应商: BOCHEM
11120 11130 11140 11115 11180 11190 11132 11122 11123 11133 11143
BOCH11120EA 0 CNY
BOCH11120 BOCH11130 BOCH11140 BOCH11115 BOCH11180 BOCH11190 BOCH11132 BOCH11122 BOCH11123 BOCH11133 BOCH11143
Laboratory jacks
实验室千斤顶
These lab jacks feature an improved scissor technology with reduced tolerances. The scissor construction is made of ¹⁸/₁₀ stainless steel.
  • Ergonomic round knob (either made of plastic or metal)
  • Smaller models up to 240×240 mm have a simple adjusting wheel which can be operated by hand
  • Larger models from 300×300 mm on should be used with a ratchet, which connects with the special adjusting wheel (ratchet is included)
  • Models with metal knob are autoclavable
DIN 12897
Certifications: DIN 12897