Dynamic climatic chamber, MK Series
供应商: BINDER GMBH
9020-0374
BIND9020-0374EA 0 CNY
BIND9020-0374
Dynamic climatic chamber, MK Series
试验箱 气候室
The MK series chambers are suitable for all heat and cold tests from –40 to 180 °C. This is an alternative to complex individual solutions for cyclical temperature tests.
  • Programmable condensation protection for test material
  • CFC-free refrigerant R-452A
  • Heated viewing window with LED interior lighting
  • Troubleshooting system with visual and audible alarms
  • Intuitive touchscreen controller with time-segment and real-time programming
  • Internal data logger, measured values can be read out in open format via USB
  • Two access ports with silicone plugs, 80 mm, left
More Product Information