Multi-channel pipettes, mechanical, variable volume, mLINE®
供应商: SARTORIUS BIOHIT
725120
BHIT725120EA 0 CNY
BHIT725120
Multi-channel pipettes, mechanical, variable volume, mLINE®
移液管和移液器 移液器
mLINE® pipettes offer a unique combination of new features for higher accuracy and precision. They include built-in replaceable tip-cone filters, which prevent pipette contamination and damage.
  • Clear and easy to read display with big numbers and small increments
  • Equipped with special spring-loaded tip cones (Optiload mechanism) allowing tip loading with equal constant force to every channel using minimum force
  • Plunger technology results in the lowest pipetting force
  • Volume range colour coding for easy pipette selection, and five colour coded caps
  • Thermal
More Product Information