Desiccator cabinets, with gas ports, dry-keeper™, SP Bel-Art
供应商: Bel-Art Products, a Part of SP
H420530002
BELAH420530002EA 0 CNY
BELAH420530002
Desiccator cabinets, with gas ports, dry-keeper™, SP Bel-Art
干燥器 干燥柜
Compact clear polystyrene desiccators, up to 5 units can be safely stacked for maximum use of laboratory bench space.
  • Pull down door slides out of the way into a compartment beneath the cabinet
  • Rear mounted gas ports are tapered to accept tubing with 6,3 to 9,5 mm
  • Gas ports can be closed to use desiccator with standard desiccant cartridges