Bags, safety pouch, SP Bel-Art
供应商: Bel-Art Products, a Part of SP
H132340000
BELAH132340000EA 0 CNY
BELAH132340000
Bags, safety pouch, SP Bel-Art
废物处理袋
Hard to pierce pouch offering protection against the danger of being cut by contaminated sharp objects. Convenient for labs, clinics and medical offices. Used with the Poxygrid® safety pouch stand for easy benchtop collection.
  • Solid paperboard pouch
  • Remains open in an oval shape by folding the bottom flaps
  • After filling, the top flaps close and lock in place for safe disposal
  • Printed with bold biohazard symbol