Desiccator cabinets, Secador®, SP Bel-Art
供应商: Bel-Art Products, a Part of SP
F420721009 F420731002 F420741002 F420731009
BELAF420721009EA 0 CNY
BELAF420721009 BELAF420731002 BELAF420741002 BELAF420731009
Desiccator cabinets, Secador®, SP Bel-Art
干燥器 干燥柜
The attractive design and sturdy construction of the Secador® range of desiccator cabinets ensures easy and reliable storage of moisture and UV sensitive products. Durastar® co-polyester construction blocks 99% of UV light and is resistant to staining, crazing and chemical attack.
  • Large doors maximise access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Rear mounted gas ports are tapered to accept tubing with ¼ to ⅜” I.D.
  • Perforated shelves facilitate circulation of dry air and have
More Product Information