Conway diffusion cell, SP Bel-Art
供应商: Bel-Art Products, a Part of SP
F409410000
BELAF409410000EA 0 CNY
BELAF409410000
Conway diffusion cell, SP Bel-Art
临床诊断系统 Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Conway diffusion cells made of smooth white molded PP with natural clear cover.
  • Obrink modification of the cell
  • Knob on the cover used to raise it
  • Circular moat of the cell receives the cover sealing off the outermost chamber
  • Steam autoclavable at 121 °C
Used for assays of carbon dioxide, ammonia, or alcohol levels in blood or serum using the diffusion method.