Test tube racks, Stack Rack™, SP Bel-Art
供应商: Bel-Art Products, a Part of SP
F188601316
BELAF188601316EA 0 CNY
BELAF188601316
Test tube racks, Stack Rack™, SP Bel-Art
架子 Tube Racks
PP, white
  • Sturdy and chemical resistant
  • Will not shatter or stain
  • With top shelf height of 70 mm
These rigid racks features interlocking end plates with handles for secure stacking when loaded with tubes up to 100 mm long. Rows are numbered and lettered for easy tube identification.