Rack for cuvettes, No-Wire™, SP Bel-Art
供应商: Bel-Art Products, a Part of SP
F185200000
BELAF185200000EA 0 CNY
BELAF185200000
Rack for cuvettes, No-Wire™, SP Bel-Art
架子 试管架
These PP rack provides gentle no-scratch support for fragile and expensive spectrophotometer cuvettes.
  • Holds forty-two 10 mm light path cuvettes
  • 6×7 row layout
  • Steam autoclavable at 121 °C