Rack for cuvettes, SP Bel-Art
供应商: Bel-Art Products, a Part of SP
F185160787 F185160000
BELAF185160787EA 0 CNY
BELAF185160787 BELAF185160000
Rack for cuvettes, SP Bel-Art
架子 试管架
Racks, made of PP, translucent. Hold fragile spectralphotometer cuvettes in complete safety.
  • Only the corners of the cuvettes touch the racks
  • Accommodate 12 cuvettes of 10 mm light path
  • Will not scratch quartz cuvettes
  • F18516-0787 features Stack Rack™ interlocking end plates for secure stacking
These two tier racks support cuvettes with a minimum of physical contact, greatly reducing the chance of damage.