Funnels, thistle
供应商: Bel-Art Products, a Part of SP
F147510000
BELAF147510000EA 0 CNY
BELAF147510000
Funnels, thistle
漏斗 过滤漏斗 Filter Funnels
PP, translucent
  • Chemical resistant
  • Unbreakable and safe
  • Steam autoclavable at 121 °C