Büchner funnels, two piece, SP Bel-Art
供应商: Bel-Art Products, a Part of SP
F146000043 H146030000 H146040000 H146010000 H146020000
BELAF146000043EA 0 CNY
BELAF146000043 BELAH146030000 BELAH146040000 BELAH146010000 BELAH146020000
Büchner funnels, two piece, SP Bel-Art
漏斗 过滤漏斗 Büchner Funnels
PP, white.
  • Autoclavable
  • Robust and lightweight
  • Top and bottom can be separated for easy cleaning
For filter paper or membrane filtration discs.