Wet surface pen, SP Bel-Art
供应商: Bel-Art Products, a Part of SP
F133820000
BELAF133820000EA 0 CNY
BELAF133820000
Wet surface pen, SP Bel-Art
Ink Pens
It can be used to correct or write on wet labels that were kept in a water bath, in liquid nitrogen or in a freezer at 0 °C. Can be also used on dry paper as a standard ball-point pen.
  • Writes clearly on wet surfaces