Micro sampling vials
供应商: Beckman Coulter
144709
BECL144709EA 0 CNY
BECL144709
Micro sampling vials
小瓶
Microvials can be used as vial inserts for small volume sample introduction into the CESI 8000, PA 800 series and P/ACE MDQ systems.
  • Microvials fit into universal vials