Collagen I culture dishes, Corning® BioCoat™
供应商: Corning
354450
BDAA354450EA 0 CNY
BDAA354450
Collagen I culture dishes, Corning® BioCoat™
培养皿 组织培养皿
PS coated with rat tail collagen type I, sterile, non-pyrogenic.
  • Manufactured in a highly controlled environment and rigorously tested to assure product consistency and performance
  • Stable at room temperature when stored in dry conditions
Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon and bone. It is an integral part of the framework that holds cells and tissues together and has been recognised as a useful matrix for improving cell culture. Used in vitro, collagen can exert effects on the adherence, morphology,More Product Information