Round-bottom polystyrene tubes, disposable, Falcon®
供应商: Corning
352001 352003 352008 352017 352025 352027 352037 352052 352054 352057 352058 352235
BDAA352001EA 0 CNY
BDAA352001 BDAA352003 BDAA352008 BDAA352017 BDAA352025 BDAA352027 BDAA352037 BDAA352052 BDAA352054 BDAA352057 BDAA352058 BDAA352235
Round-bottom polystyrene tubes, disposable, Falcon®
管子 培养管
These disposable, clear polystyrene tubes are best suited for procedures requiring high optical clarity, and thermal and chemical stability. Provide reliable containment of laboratory fluid samples.
  • Dual position snap-cap prevents sample loss with ease of handling, heavier gauge walls, and unique construction provide a secure, positive seal
  • Medical-style packaging materials
  • Tube dimensions and volumes are approximate
  • Widely used and referenced in laboratory protocols
  • Sterile, nonpyrogenicity tested to less than 0,1 EU/ml (Cat. No. 352008 nonsterile)