Cryogenic vials, TruCool®
供应商: BIOCISION
BCS-2510 BCS-2511 BCS-2512 BCS-2513 BCS-2514 BCS-2515 BCS-2516 BCS-2517 BCS-2502 BCS-2501 BCS-2503 BCS-2504 BCS-2505
BCISBCS-2510EA 0 CNY
BCISBCS-2510 BCISBCS-2511 BCISBCS-2512 BCISBCS-2513 BCISBCS-2514 BCISBCS-2515 BCISBCS-2516 BCISBCS-2517 BCISBCS-2502 BCISBCS-2501 BCISBCS-2503 BCISBCS-2504 BCISBCS-2505
Cryogenic vials, TruCool®
小瓶
TruCool® leak-proof cryogenic vials feature a screw cap with a thermally-fused gasket to ensure a tight seal. The gasket is a co-molded thermoplastic elastomer (TPE) layer, which prevents leaking, slipping and risk of contamination. The gasket is 95 kPa certified to provide a leak-proof seal. These vials are sterile and individually barcoded with a unique identifier which can be read with common barcode readers. Ideal for use in CoolRack® CF cryogenic vial modules and CoolCell® alcohol-free cell freezing containers. Recommended for use in vapor phase LN₂ only.
Packaging: CasesMore Product Information