Ultrapure water systems, Barnstead™ MicroPure™
供应商: Thermo Fisher Scientific
50132370 50132372 50132373 50132374
BARN50132370EA 0 CNY
BARN50132370 BARN50132372 BARN50132373 BARN50132374
Ultrapure water systems, Barnstead™ MicroPure™
超纯水系统 纯水和超纯水系统
These water purification systems deliver ultrapure 18,2 MΩ.cm water with consistent quality. They are designed for laboratories that require up to 15 L/day of ultrapure water for HPLC or other instrumental analyses, sample preparation, glassware rinsing, or molecular biology work in which endotoxins and/or nucleases would interfere.
  • Optional integrated 6 L feed water reservoir: System can be placed without direct access to a water line
  • Validated 0,2 µM final filter can be autoclaved up to 5 times
  • Standard feed water monitor supports consistent water
More Product Information
现在订购
规格
型号 UV/UF UV/UF, with tank UV UV, with tank
Environmentally Preferable
给水 RO, DI or distilled water RO, DI or distilled water RO, DI or distilled water RO, DI or distilled water
流速 1,0 l/min 0,8 l/min 1,5 l/min 1,0 l/min
电导率 0,055 µS/cm 0,055 µS/cm 0,055 µS/cm 0,055 µS/cm
TOC content 1 - 5 ppb 1 - 5 ppb 1 - 5 ppb 1 - 5 ppb
Resistivity at +25 °C 18,2 MΩ.cm 18,2 MΩ.cm 18,2 MΩ.cm 18,2 MΩ.cm
细菌 <1 CFU/ml <1 CFU/ml <1 CFU/ml <1 CFU/ml
宽度×深度×高度 305×300×545 mm 305×390×545 mm 305×300×545 mm 305×390×545 mm
重量 15 kg 17 kg 15 kg 17 kg