乙酸异丙酯 ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®

供应商: Avantor
Danger

Synonyms: Acetic acid isopropyl ester

Management of Change (MOC) category = N

BAKRU385-03EA 1868 CNY
BAKRU385-03 BAKRU385-07 BAKRU385-26 BAKRU385-6K
乙酸异丙酯 ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
乙酸异丙酯
分子式: C₅H₁₀O₂
分子量: 102.13 g/mol
沸点: 85 °C (1013 hPa)
溶点: –62 °C
密度: 0.8732 g/cm³ (20 °C)
闪点: 4 °C
存储条件: Ambient
MDL编号: MFCD00008877
CAS号: 108-21-4
EINECS: 203-561-1
UN号: 1220
ADR: 3 II
Merck Index: 13,05225

现在订购

产品技术规格

For Laboratory,Research,or Manufacturing Use
Assay (by GC) ≥99.0 %
Density (g/mL) at 25°C 0.86 - 0.87
Residue after Evaporation ≤0.0050 %
Acidity (as CH₃COOH) ≤0.01 %
Water (H₂O) ≤0.1 %

规格

Environmentally Preferable Safer Human and Environmental Health
环境认证 Chem21 Solvent Selection Guide recommended by default and after discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR