, BAKER ANALYZED® LC-MS, J.T.Baker®

供应商: Avantor
Danger

BAKR9832-03EA 12248 CNY
BAKR9832-03 BAKR9832-02
, BAKER ANALYZED® LC-MS, J.T.Baker®
Acetonitrile with 0,1% (v/v) formic acid

Clear, colorless liquid. For use in liquid chromatography and mass spectrometry.

存储条件: Ambient
EINECS: 200-579-1
UN号: 1993
ADR: 3 II

现在订购

产品技术规格

For Laboratory,Research,or Manufacturing Use Passes Test
Appearance Passes Test
Formic Acid 0.095 - 0.105 %
Gradient Elution Test (a.u.) - 254 nm ≤ 0.01
Residue after Evaporation ≤ 1.0 ppm
Water (H₂O) ≤ 200 ppm
LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine) ≤ 50 ppb
Trace Impurities - Aluminum (Al) ≤ 50.0 ppb
Trace Impurities - * Calcium (Ca) ≤ 50.0 ppb
Trace Impurities - Iron (Fe) ≤ 50.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 50.0 ppb
Trace Impurities - Potassium (K) ≤ 50 ppb
Trace Impurities - * Sodium (Na) ≤ 50.0 ppb
* May change over time due to extraction from glass container.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR