硫酸 ≥ 95% 95,0-98,0 Acidimetry NF, FCC, J.T.Baker®

供应商: Avantor
Danger

BAKR9675-02EA 0 CNY
BAKR9675-02 BAKR9675-03 BAKR9675-R BAKR9675-07 BAKR9675-08 BAKR9675-25
硫酸 ≥ 95% 95,0-98,0 Acidimetry NF, FCC, J.T.Baker®
硫酸
分子式: H₂SO₄
分子量: 98.08 g/mol
沸点: ∼335 °C (1013 hPa)
溶点: 3 °C
密度: 1.84 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00064589
CAS号: 7664-93-9
EINECS: 231-639-5
UN号: 1830
ADR: 8 II
Merck Index: 14,08974

现在订购

产品技术规格

Meets N.F. Requirements -
Meets F.C.C. Requirements -
GMP Manufactured Product -
Food GMP Manufactured Product -
CAUTION: For Manufacturing, processing or repackaging 95.0 - 98.0 %
Bulk Pharmaceutical Chemical ≤ 1 ppm
Bulk Food Chemical ≤ 0.005 %
NF - Assay (H₂SO₄) (by acidimetry) 95.0 - 98.0 %
NF - Arsenic (As) ≤1 ppm
NF - Chloride (Cl) ≤0.005 %
NF - Identification Passes Test
NF - Reducing Substances Passes Test
NF - Residue on Ignition ≤0.005 %
FCC - Assay (H₂SO₄) (by acidimetry) 95.0 - 98.0 %
FCC - Arsenic (As) ≤1 ppm
FCC - Chloride (Cl) ≤0.005 %
FCC - Iron (Fe) ≤0.02 %
FCC - Identification Passes Test
FCC - Lead (Pb) ≤5 mg/kg
FCC - Nitrate (NO₃) ≤10 mg/kg
FCC - Reducing Substances Passes Test
FCC - Selenium (Se) ≤0.002 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR