乙酸 glacial ≥99.7%, CMOS for microelectronic, J.T.Baker®

供应商: Avantor
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

BAKR9513-07EA 0 CNY
BAKR9513-07
乙酸 glacial ≥99.7%, CMOS for microelectronic, J.T.Baker®
乙酸
分子式: H₃CCOOH
分子量: 60.05 g/mol
沸点: 118
溶点: 17
密度: 1.05 g/cm³ (20 °C)
闪点: 38.5
存储条件: Ambient
MDL编号: MFCD00036152
CAS号: 64-19-7
EINECS: 200-580-7
UN号: 2789
ADR: 8 II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

现在订购

产品技术规格

For Microelectronic Use
IMPORTANT: Material will freeze if stored below 17 °C (63°F).
Assay (CH₃COOH) (by freezing point) ≥ 99.7 %
Color (APHA) ≤ 10
Acetic Anhydride ((CH₃CO)₂O) ≤ 0.01 %
Acetaldehyde ≤ 0.05 %
Residue after Evaporation ≤ 4 ppm
Solubility in H₂O Passes Test
Specific Gravity at 20°/20°C ≥ 1.048
Substances Reducing Dichromate Passes Test
Substances Reducing Permanganate Passes Test
Chloride (Cl) ≤ 1 ppm
Phosphate (PO₄) ≤ 1 ppm
Sulfate (SO₄) ≤ 0.5 ppm
Trace Impurities - Aluminum (Al) ≤ 50.0 ppb
Arsenic and Antimony (as As) ≤ 5.0 ppb
Trace Impurities - Barium (Ba) ≤ 10.0 ppb
Trace Impurities - Beryllium (Be) ≤ 10.0 ppb
Trace Impurities - Bismuth (Bi) ≤ 50 ppb
Trace Impurities - Boron (B) ≤ 10.0 ppb
Trace Impurities - Cadmium (Cd) ≤ 10.0 ppb
Trace Impurities - Calcium (Ca) ≤ 200.0 ppb
Trace Impurities - Chromium (Cr) ≤ 30.0 ppb
Trace Impurities - Cobalt (Co) ≤ 10.0 ppb
Trace Impurities - Copper (Cu) ≤ 20.0 ppb
Trace Impurities - Gallium (Ga) ≤ 10.0 ppb
Trace Impurities - Germanium (Ge) ≤ 10.0 ppb
Trace Impurities - Gold (Au) ≤ 10.0 ppb
Heavy Metals (as Pb) ≤ 300.0 ppb
Trace Impurities - Iron (Fe) ≤ 100.0 ppb
Trace Impurities - Lead (Pb) ≤ 100 ppb
Trace Impurities - Lithium (Li) ≤ 10.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 50.0 ppb
Trace Impurities - Manganese (Mn) ≤ 10.0 ppb
Trace Impurities - Molybdenum (Mo) ≤ 10.0 ppb
Trace Impurities - Nickel (Ni) ≤ 25.0 ppb
Trace Impurities - Niobium (Nb) ≤ 10.0 ppb
Trace Impurities - Potassium (K) ≤ 100 ppb
Trace Impurities - Silicon (Si) ≤ 50 ppb
Trace Impurities - Silver (Ag) ≤ 10.0 ppb
Trace Impurities - Sodium (Na) ≤ 100.0 ppb
Trace Impurities - Strontium (Sr) ≤ 10.0 ppb
Trace Impurities - Tantalum (Ta) ≤ 10.0 ppb
Trace Impurities - Thallium (Tl) ≤ 50.0 ppb
Trace Impurities - Tin (Sn) ≤ 50 ppb
Trace Impurities - Titanium (Ti) ≤ 100.0 ppb
Trace Impurities - Vanadium (V) ≤ 10.0 ppb
Trace Impurities - Zinc (Zn) ≤ 50 ppb
Trace Impurities - Zirconium (Zr) ≤ 10.0 ppb
For additional information, go to www.askavantor.com. Search keywords
"freezing" and product name.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR