丙酮 ≥99.4% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®

供应商: Avantor
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

Packaged under inert gas.

BAKR9254-03EA 2119 CNY
BAKR9254-03 BAKR9254-02 BAKR9254-3P BAKR9254-50 BAKR9254-7K BAKR9254-95 BAKR9254-98 BAKR9254-99 BAKR9254-9D BAKR9254-9E BAKR9254-9H BAKR9254-H1 BAKR9254-Q3 BAKR9254-V8 BAKR9254-WA BAKR9254-Y6 BAKR9254-YQ BAKR9254-YV BAKR9254-ZC
丙酮 ≥99.4% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
丙酮

BAKER ULTRA RESI-ANALYZED® Reagent—ultra high-purity solvents and salts developed for organic residue extraction/concentration procedures.


These solvents are fully characterised and lot controlled by ECD, FID, or other method-specific detectors. Advanced stabiliser systems are incorporated to ensure solvent stability and superior performance. Solvents are suitable for exacting EPA sample cleanup protocols.

分子式: CH₃COCH₃
分子量: 58.08 g/mol
沸点: 56.2
溶点: –95.4 °C
密度: 0.792 g/cm³ (20 °C)
闪点: <–20 °C (closed cup)
存储条件: Ambient
MDL编号: MFCD00008765
CAS号: 67-64-1
EINECS: 200-662-2
UN号: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

现在订购

产品技术规格

For Laboratory,Research,or Manufacturing Use Min. 99.4 %
Assay ((CH₃)₂CO) (by GC, corrected for water) ≥99.4 %
Color (APHA) ≤10
Residue after Evaporation ≤1.0 ppm
Substances Reducing Permanganate Passes Test
Titrable Acid (µeq/g) ≤0.3
Titrable Base (µeq/g) ≤0.6
Water (H₂O) ≤0.5 %
FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤5
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
MEETS SPECIFICATIONS WITHIN THE EXPIRATION PERIOD

规格

Environmentally Preferable Safer Human and Environmental Health
环境认证 Chem21 Solvent Selection Guide recommended after discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR