J.T.Baker® BAKERBOND®, Ion Exchange Chromatography Resin, PolyQUAT Multimode
供应商: Avantor
BAKR7603-01EA 0 CNY
BAKR7603-01 BAKR7603-02 BAKR7603-03 BAKR7603-04 BAKR7603-05
J.T.Baker® BAKERBOND®, Ion Exchange Chromatography Resin, PolyQUAT Multimode
色谱吸附剂 高效液相色谱吸附剂
The J.T.Baker® BAKERBOND® PolyQUAT multimode ion exchange (IEX) chromatography resin functions as a strong anion exchange resin with improved selectivity for superior separation in process chromatography applications.
Powered by a proprietary surface chemistry combining strong anion exchange functionality with weak anionic exchange sites and hydrophilicity, BAKERBOND® PolyQUAT multimode cation exchange chromatography media utilizes its unique selectivity to enable higher purification performance across a wide range of biopharmaceuticals.
Designed and manufactured by Avantor to the high standards established for our J.T.Baker® brand chemicals, BAKERBOND® PolyQUAT ion exchange resin can be used with other J.T.Baker® BAKERBOND® resins to enhance the separation power of your chromatography process steps.
订购须知 1 ml and 5 ml lab columns are also available under complementary products.
现在订购
规格
Environmentally Preferable
工作 pH 2-14