J.T.Baker® BAKERBOND® Q2100, New Generation UHPLC and HPLC Columns
供应商: Avantor
BAKR7358-14EA 5203 CNY
BAKR7358-14 BAKR7395-52 BAKR7342-52 BAKR7351-52 BAKR7395-12 BAKR7342-12 BAKR7351-12 BAKR7405-52 BAKR7354-52 BAKR7358-52 BAKR7373-52 BAKR7405-32 BAKR7354-32 BAKR7358-32 BAKR7373-32 BAKR7405-42 BAKR7354-42 BAKR7358-42 BAKR7373-42 BAKR7405-54 BAKR7354-54 BAKR7358-54 BAKR7373-54 BAKR7405-34 BAKR7354-34 BAKR7358-34 BAKR7373-34 BAKR7405-14 BAKR7354-14 BAKR7375-42 BAKR7411-22 BAKR7378-22 BAKR7382-22 BAKR7375-22 BAKR7411-14 BAKR7378-14 BAKR7382-14 BAKR7375-14 BAKR7411-24 BAKR7378-24 BAKR7382-24 BAKR7375-24 BAKR7373-14 BAKR7411-42 BAKR7378-42 BAKR7382-42 BAKR7411-2H BAKR7411-4H BAKR7378-2H BAKR7378-4H BAKR7382-2H BAKR7382-4H BAKR7375-2H BAKR7375-4H BAKR7395-2U BAKR7342-2U BAKR7351-2U BAKR7354-24 BAKR7354-44 BAKR7358-24 BAKR7358-44 BAKR7373-24 BAKR7373-44 BAKR7375-44 BAKR7378-44 BAKR7382-44 BAKR7405-24 BAKR7405-44 BAKR7411-44 BAKR7405-64 BAKR7354-64 BAKR7382-26
J.T.Baker® BAKERBOND® Q2100, New Generation UHPLC and HPLC Columns
色谱柱
BAKERBOND® Q2100 LC phases have been specially designed to solve the challenges of most routine samples. They can also be a good starting point for new method development projects. With options of four widely used chemistries and three particle sizes BAKERBOND® Q2100 can be a useful addition to your analytical toolkit.
  • High efficiency UHPLC/HPLC columns
  • Based on high purity spherical silica
  • C18, C8, phenyl-hexyl and silica phases
  • Available with 1,8, 3, 5 µm particle sizes